missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHOSPHO2 Protein

Product Code. 15902339
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15902339 10 μg 10µg
15922339 2 μg 2µg
15912339 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15902339 Supplier enQuireBio™ Supplier No. QP1304410ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PHOSPHO2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PHOSPHO2 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSHMKILLV FDFDNTIIDD NSDTWIVQCA PNKKLPIELR DSYRKGFWTE FMGRVFKYLG DKGVREHEMK RAVTSLPFTP GMVELFNFIR KNKDKFDCII ISDSNSVFID WVLEAASFHD IFDKVFTNPA AFNSNGHLTV ENYHTHSCNR CPKNLCKKVV LIEFVDKQLQ QGVNYTQIVY IGDGGNDVCP VTFLKNDDVA MPRKGYTLQK TLSRMSQNLE PMEYSVVVWS SGVDIISHLQ FLIKD
Buffer The PHOSPHO2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.