missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Peroxisomal bifunctional enzyme Protein

A DNA sequence encoding the Homo sapiens (Human) Peroxisomal bifunctional enzyme, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7513-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049120

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001159887.1
Peroxisomal bifunctional enzyme Protein
Research Use Only
EHHADH
Recombinant Protein
E. coli
MAEYTRLHNA LALIRLRNPP VNAISTTLLR DIKEGLQKAV IDHTIKAIVI CGAEGKFSAG ADIRGFSAPR TFGLTLGHVV DEIQRNEKPV VAAIQGMAFG GGLELALGCH YRIAHAEAQV GLPEVTLGLL PGARGTQLLP RLTGVPAALD LITSGRRILA DEALKLGILD KVVNSDPVEE AIRFAQRVSD QPLESRRLCN KPIQSLPNMD SIFSEALLKM RRQHPGCLAQ EACVRAVQAA VQYPYEVGIK KEEELFLYLL QSGQARALQY AFFAERKANK WSTPSGASWK TASARPVSSV GVVGLGTMGR GIVISFARAR IPVIAVDSDK NQLATANKMI TSVLEKEASK MQQSGHPWSG PKPRLTSSVK ELGGVDLVIE AVFEEMSLKK QVFAELSAVC KPEAFLCTNT SALDVDEIAS STDRPHLVIG THFFSPAHVM KLLEVIPSQY SSPTTIATVM NLSKKIKKIG VVVGNCFGFV GNRMLNPYYN QAYFLLEEGS KPEEVDQVLE EFGFKMGPFR VSDLAGLDVG WKSRKGQGLT GPTLLPGTPA RKRGNRRYCP IPDVLCELGR FGQKTGKGWY QYDKPLGRIH KPDPWLSKFL SRYRKTHHIE PRTISQDEIL ERCLYSLINE AFRILGEGIA ASPEHIDVVY LHGYGWPRHK GGPMFYASTV GLPTVLEKLQ KYYRQNPDIP QLEPSDYLKK LASQGNPPLK EWQSLAGSPS SKL
Greater than 90% as determined by SDS-PAGE.
1962
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions