missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAX9 Protein
A cDNA sequence encoding the PAX9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12970-1mg
Additional Details : Weight : 0.01000kg
Specifications
PAX9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPAFGE VNQLGGVFVN GRPLPNAIRL RIVELAQLGI RPCDISRQLR VSHGCVSKIL ARYNETGSIL PGAIGGSKPR VTTPTVVKHI RTYKQRDPGI FAWEIRDRLL ADGVCDKYNV PSVSSISRIL RNKIGNLAQQ GHYDSYKQHQ PTPQPALPYN HIYSYPSPIT AAAAKVPTPP GVPAIPGSVA MPRTWPSSHS VTDILGIRSI TDQVSDSSPY HSPKVEEWSS LGRNNFPAAA PHAVNGLEKG ALEQEAKYGQ APNGLPAVGS FVSASSMAPY PTPAQVSPYM TYSAAPSGYV AGHGWQHAGG TSLSPHNCDI PASLAFKGMQ AAREGSHSVT ASAL | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PAX9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |