missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Orexin receptor type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Orexin receptor type 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9175-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001517.2 | |
Orexin receptor type 2 Protein | |
Research Use Only | |
HCRTR2 | |
Recombinant Protein | |
Yeast | |
MSGTKLEDSP PCRNWSSASE LNETQEPFLN PTDYDDEEFL RYLWREYLHP KEYEWVLIAG YIIVFVVALI GNVLV | |
Greater than 90% as determined by SDS-PAGE. |
3062 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |