missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein

Product Code. 18187190
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18187190

Brand: R&D Systems™ 377HB050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein is derived from E. coli. The Recombinant Human NRG1-beta 1/HRG1-beta 1 ECD Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 3084
Molecular Weight (g/mol) 26.9 kDa
Name Neuregulin-1 beta 1/NRG1 beta 1 Extracellular Domain
Quantity 50 μg
Source E. coli-derived human Neuregulin-1 beta 1/NRG1 beta 1 protein Ser2-Lys246
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.