missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurotensin Protein
A DNA sequence encoding the Homo sapiens (Human) Neurotensin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6438-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006174.1 | |
Neurotensin Protein | |
Research Use Only | |
NTS | |
Recombinant Protein | |
E. coli | |
SDSEEEMKAL EADFLTNMHT SKISKAHVPS WKMTLLNVCS LVNNLNSPAE ETGEVHEEEL VARRKLPTAL DGFSLEAMLT IYQLHKICHS RAFQHWELIQ EDILDTGNDK NGKEEVIKRK | |
Greater than 90% as determined by SDS-PAGE. |
4922 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |