missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neuronal calcium sensor 1 Protein

Product Code. 15989659
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15989659 100 μg 100µg
15999659 10 μg 10µg
15939669 50 μg 50µg
15919669 200 μg 200µg
15929669 500 μg 500µg
15909669 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15989659 Supplier enQuireBio™ Supplier No. QP6056ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Neuronal calcium sensor 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001122298.1
Gene ID (Entrez) 23413
Name Neuronal calcium sensor 1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NCS1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence GKSNSKLKPE VVEELTRKTY FTEKEVQQWY KGFIKDCPSG QLDAAGFQKI YKQFFPFGDP TKFATFVFNV FDENKDGRIE FSEFIQALSV TSRGTLDEKL RWAFKLYDLD NDGYITRNEM LDIVDAIYQM VGNTVELPEE ENTPEKRVDR IFAMMDKNAD GKLTLQEFQE GSKADPSIVQ ALSLYDGLV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.