missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFA2 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8432-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171941.1 | |
NDUFA2 Protein | |
Research Use Only | |
NDUFA2 | |
Recombinant Protein | |
E. coli | |
AAASRGVGAK LGLREIRIHL CQRSPGSQGV RDFIEKRYVE LKKANPDLPI LIRECSDVQP KLWARYAFGQ ETNVPLNNFS ADQVTRALEN VLSGKA | |
Greater than 90% as determined by SDS-PAGE. |
4695 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |