missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NAD-dependent malic enzyme, mitochondrial Protein

Product Code. 15972918
Change view
Click to view available options
Quantity:
100 μg
25 μg
250 μg
5 μg
Unit Size:
100µg
250µg
25µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15972918 100 μg 100µg
15902928 5 μg 5µg
15992918 25 μg 25µg
15982918 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15972918 Supplier enQuireBio™ Supplier No. QP10156ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human NAD-dependent malic enzyme, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001161807.1
Gene ID (Entrez) 4200
Name NAD-dependent malic enzyme, mitochondrial Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol ME2
Protein Length Partial (see sequence information for more details).
Biological Activity Malic Enzyme activity was assayed spectrophotometrically at 340nm as described in Mandela and Sauer (1975). The standard reaction mixture contained 50 mM Tris-HCl, 3 mM MnCl2, 5 mM malate, 0.12 mM NADP+, 2.5 mM fumarate. Assay was performed in a Beckman spectrophotometer. The Km value is 1.5 ± 0.6 mM.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MLHIKEKGKP LMLNPRTNKG MAFTLQERQM LGLQGLLPPK IETQDIQALR FHRNLKKMTS PLEKYIYIM GIQERNEKLF YRILQDDIES LMPIVYTPTV GLACSQYGHI FRRPKGLFIS ISDRGHVRS IVDNWPENHV KAVVVTDGER ILGLGDLGVY GMGIPVGKLC LYTACAGIRP DRCLPVCID VGTDNIALLK DPFYMGLYQK RDRTQQYDDL IDEFMKAITD RYGRNTLIQF EDFGNHNAF RFLRKYREKY CTFNDDIQGT AAVALAGLLA AQKVISKPIS EHKILFLGAG EAALGIANL IVMSMVENGL SEQEAQKKIW MFDKYGLLVK GRKAKIDSYQ EPFTHSAPES IPDTFEDAV NILKPSTIIG VAGAGRLFTP DVIRAMASIN ERPVIFALSN PTAQAECTAE EAYTLTEGR CLFASGSPFG PVKLTDGRVF TPGQGNNVYI FPGVALAVIL CNTRHISDSV FLEAAKALT SQLTDEELAQ GRLYPPLANI QEVSINIAIK VTEYLYANKM AFRYPEPEDK AKYVKERTW RSEYDSLLPD VYEWPESASS PPVITEHHHH HH>
Buffer 0.2um filtered PBS, pH 7.4. , lyophilized
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.