missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human N-acetylmuramoyl-L-alanine amidase Protein

A DNA sequence encoding the Homo sapiens (Human) N-acetylmuramoyl-L-alanine amidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7947-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051781

  • 1740.00 € / 500µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_443122.3
N-acetylmuramoyl-L-alanine amidase Protein
Research Use Only
PGLYRP2
Recombinant Protein
Yeast
SLPLLMDSVI QALAELEQKV PAAKTRHTAS AWLMSAPNSG PHNRLYHFLL GAWSLNATEL DPCPLSPELL GLTKEVARHD VREGKEYGVV LAPDGSTVAV EPLLAGLEAG LQGRRVINLP LDSMAAPWET GDTFPDVVAI APDVRATSSP GLRDGSPDVT TADIGANTPD ATKGCPDVQA SLPDAKAKSP PTMVDSLLAV TLAGNLGLTF LRGSQTQSHP DLGTEGCWDQ LSAPRTFTLL DPKASLLTMA FLNGALDGVI LGDYLSRTPE PRPSLSHLLS QYYGAGVARD PGFRSNFRRQ NGAALTSASI LAQQVWGTLV LLQRLEPVHL QLQCMSQEQL AQVAANATKE FTEAFLGCPA IHPRCRWGAA PYRGRPKLLQ LPLGFLYVHH TYVPAPPCTD FTRCAANMRS MQRYHQDTQG WGDIGYSFVV GSDGYVYEGR GWHWVGAHTL GHNSRGFGVA IVGNYTAALP TEAALRTVRD TLPSCAVRAG LLRPDYALLG HRQLVRTDCP GDALFDLLRT WPHFTATVKP RPARSVSKRS RREPPPRTLP ATDLQ
Greater than 90% as determined by SDS-PAGE.
114770
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions