missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myotubularin-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Myotubularin-related protein 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8264-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004677.3 | |
Myotubularin-related protein 1 | |
Research Use Only | |
MTMR7 | |
Recombinant Protein | |
E. coli | |
MEHIRTPKVE NVRLVDRVSP KKAALGTLYL TATHVIFVEN SPDPRKETWI LHSQISTIEK QATTATGCPL LIRCKNFQII QLIIPQERDC HDVYISLIRL ARPVKYEELY CFSFNPMLDK EEREQGWVLI DLSEEYTRMG LPNHYWQLSD VNRDYRVCDS YPTELYVPKS ATAHIIVGSS KFRSRRRFPV LSYYYKDNHA SICRSSQPLS GFSARCLEDE QMLQAIRKAN PGSDFVYVVD TRPKLNAMAN RAAGKGYENE DNYSNIKFQF IGIENIHVMR NSLQKMLEVC ELKSPSMSDF LWGLENSGWL RHIKAIMDAG IFIAKAVSEE GASVLVHCSD GWDRTAQVCS VASLLLDPHY RTLKGFMVLI EKDWISFGHK FNHRYGNLDG DPKEISPVID QFIECVWQLM EQFPCAFEFN ERFLIHIQHH IYSCQFGNFL CNSQKERREL KIQERTYSLW AHLWKNRADY LNPLFRADHS QTQGTLHLPT TPCNFMYKFW SGMYNRFEKG MQPRQSVTDY LMAVKEETQQ LEEELEALEE RLEKIQKVQL NCTKVKSKQS EPSKHSGFST SDNSIANTPQ DYSGNMKSFP SRSPSQGDED SALILTQDNL KSSDPDLSAN SDQESGVEDL SCRSPSGGEH APSEDSGKDR DSDEAVFLTA | |
Greater than 90% as determined by SDS-PAGE. |
9108 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |