missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-binding protein C, cardiac-type Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-binding protein C, cardiac-type, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7532-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000247.2 | |
Myosin-binding protein C, cardiac-type Protein | |
Research Use Only | |
Mybpc3 | |
Recombinant Protein | |
E. coli | |
MPEPGKKPVS AFSKKPRSVE VAAGSPAVFE AETERAGVKV RWQRGGSDIS ASNKYGLATE GTRHTLTVRE VGPADQGSYA VIAGSSKVKF DLKVIEAEKA EPMLAPAPAP AEATGAPGEA PAPAAELGES APSPKGSSSA ALNGPTPGAP DDPIGLFVMR PQDGEVTVGG SITFSARVAG ASLLKPPVVK WFKGKWVDLS SKVGQHLQLH DSYDRASKVY LFELHITDAQ PAFTGSYRCE VSTKDKFDCS NFNLTVHEAM GTGDLDLLSA FRRTSLAGGG RRISDSHEDT GILDFSSLLK KRDSFRTPRD SKLEAPAEED VWEILRQA | |
Greater than 90% as determined by SDS-PAGE. |
4607 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |