Learn More
enQuireBio™ Recombinant Human MESDC1 Protein
A cDNA sequence encoding the MESDC1 was constructed and used to recombinantly synthesize the protein.
137.00 € - 4090.00 €
Specifications
Name | MESDC1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911209
|
enQuireBio™
QP12680-5UG |
5 μg |
137.00 €
5µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901209
|
enQuireBio™
QP12680-20UG |
20 μg |
212.00 €
20µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991199
|
enQuireBio™
QP12680-1MG |
1 mg |
4090.00 €
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MESDC1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MESDC1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASGSAG KPTGEAASPA PASAIGGASS QPRKRLVSVC DHCKGKMQLV ADLLLLSSEA RPVLFEGPAS SGAGAESFEQ CRDTIIARTK GLSILTHDVQ SQLNMGRFGE AGDSLVELGD LVVSLTECSA HAAYLAAVAT PGAQPAQPGL VDRYRVTRCR HEVEQGCAVL RATPLADMTP QLLLEVSQGL SRNLKFLTDA CALASDKSRD RFSREQFKLG VKCMSTSASA LLACVREVKV APSELARSRC ALFSGPLVQA VSALVGFATE PQFLGRAAAV SAEGKAVQTA ILGGAMSVVS ACVLLTQCLR DLAQHPDGGA KMSDHRERLR NSACAVSEGC TLLSQALRER SSPRTLPPVN SNSVN | |
Greater than 95.0% as determined by SDS-PAGE. |