missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MART-1 / MLANA Protein

A DNA sequence encoding the Homo sapiens (Human) MART-1 / MLANA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7586-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069570

  • 2385.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005502.1
MART-1 / MLANA Protein
Research Use Only
MLANA
Recombinant Protein
E. coli
MPREDAHFIY GYPKKGHGHS YTTAEEAAGI GILTVILGVL LLIGCWYCRR RNGYRALMDK SLHVGTQCAL TRRCPQEGFD HRDSKVSLQE KNCEPVVPNA PPAYEKLSAE QSPPPYSP
Greater than 90% as determined by SDS-PAGE.
2315
1 mg
Not determined.
Full Length
Human
His-B2M
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions