missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lymphocyte antigen 6E Protein
A DNA sequence encoding the Homo sapiens (Human) Lymphocyte antigen 6E, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001120685.1 |
---|---|
Gene ID (Entrez) | 4061 |
Name | Lymphocyte antigen 6E Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069430
|
enQuireBio™
QP7565-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089430
|
enQuireBio™
QP7565-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079430
|
enQuireBio™
QP7565-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099430
|
enQuireBio™
QP7565-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009440
|
enQuireBio™
QP7565-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019440
|
enQuireBio™
QP7565-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_001120685.1 | |
Lymphocyte antigen 6E Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4061 | |
Research Use Only | |
LY6E | |
Recombinant Protein | |
E. coli | |
LMCFSCLNQK SNLYCLKPTI CSDQDNYCVT VSASAGIGNL VTFGHSLSKT CSPACPIPEG VNVGVASMGI SCCQSFLCNF S | |
Greater than 90% as determined by SDS-PAGE. |