missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LTBR / TNFRSF3 Protein
A DNA sequence encoding the Homo sapiens (Human) LTBR / TNFRSF3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8595-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257916.1 | |
LTBR / TNFRSF3 Protein | |
Research Use Only | |
LTBR | |
Recombinant Protein | |
E. coli | |
QAVPPYASEN QTCRDQEKEY YEPQHRICCS RCPPGTYVSA KCSRIRDTVC ATCAENSYNE HWNYLTICQL CRPCDPVMGL EEIAPCTSKR KTQCRCQPGM FCAAWALECT HCELLSDCPP GTEAELKDEV GKGNNHCVPC KAGHFQNTSS PSARCQPHTR CENQGLVEAA PGTAQSDTTC KNPLEPLPPE MSGT | |
Greater than 90% as determined by SDS-PAGE. |
4055 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |