missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LTBR / TNFRSF3 Protein

A DNA sequence encoding the Homo sapiens (Human) LTBR / TNFRSF3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8595-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065481

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001257916.1
LTBR / TNFRSF3 Protein
Research Use Only
LTBR
Recombinant Protein
E. coli
QAVPPYASEN QTCRDQEKEY YEPQHRICCS RCPPGTYVSA KCSRIRDTVC ATCAENSYNE HWNYLTICQL CRPCDPVMGL EEIAPCTSKR KTQCRCQPGM FCAAWALECT HCELLSDCPP GTEAELKDEV GKGNNHCVPC KAGHFQNTSS PSARCQPHTR CENQGLVEAA PGTAQSDTTC KNPLEPLPPE MSGT
Greater than 90% as determined by SDS-PAGE.
4055
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions