missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LITAF Protein
A DNA sequence encoding the Homo sapiens (Human) LITAF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6312-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129944.1 | |
LITAF Protein | |
Research Use Only | |
LITAF | |
Recombinant Protein | |
E. coli | |
MSVPGPYQAA TGPSSAPSAP PSYEETVAVN SYYPTPPAPM PGPTTGLVTG PDGKGMNPPS YYTQPAPIPN NNPITVQTVY VQHPITFLDR PIQMCCPSCN KMIVSQLSYN AGALTWLSCG SLCLLGCIAG CCFIPFCVDA LQDVDHYCPN CRALLGTYKR L | |
Greater than 80% as determined by SDS-PAGE. |
9516 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |