missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LITAF Protein

A DNA sequence encoding the Homo sapiens (Human) LITAF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6312-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001129944.1
LITAF Protein
Research Use Only
LITAF
Recombinant Protein
E. coli
MSVPGPYQAA TGPSSAPSAP PSYEETVAVN SYYPTPPAPM PGPTTGLVTG PDGKGMNPPS YYTQPAPIPN NNPITVQTVY VQHPITFLDR PIQMCCPSCN KMIVSQLSYN AGALTWLSCG SLCLLGCIAG CCFIPFCVDA LQDVDHYCPN CRALLGTYKR L
Greater than 80% as determined by SDS-PAGE.
9516
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions