missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human La-related protein 1B Protein
A DNA sequence encoding the Homo sapiens (Human) La-related protein 1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7726-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265533.1 | |
La-related protein 1B Protein | |
Research Use Only | |
LARP1B | |
Recombinant Protein | |
E. coli | |
MDSRDRGPGT SSVSTSNASP SEGAPLAGSY GCTPHSFPKF QHPSHELLKE NGFTQQVYHK YRRRCLSERK RLGIGQSQEM NTLFRFWSFF LRDHFNKKMY EEFRQLAWED AKENYRYGLE CLFRFYSYGL EKKFRREIFQ DFQEETKKDY ESGQLYGLEK FWAYLKYSQS KTQSIDPKLQ EYLCSFKRLE DFRVDEADPI E | |
Greater than 90% as determined by SDS-PAGE. |
55132 | |
10 μg | |
Not determined. | |
Full Length of Isoform 7 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |