missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kynurenine--oxoglutarate transaminase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Kynurenine--oxoglutarate transaminase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7538-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001116143.1 | |
Kynurenine--oxoglutarate transaminase 1 Protein | |
Research Use Only | |
CCBL1 | |
Recombinant Protein | |
E. coli | |
MAKQLQARRL DGIDYNPWVE FVKLASEHDV VNLGQGFPDF PPPDFAVEAF QHAVSGDFML NQYTKTFVII IEPFFDCYEP MTMMAGGRPV FVSLKPGPIQ NGELGSSSNW QLDPMELAGK FTSRTKALVL NTPNNPLGKV FSREELELVA SLCQQHDVVC ITDEVYQWMV YDGHQHISIA SLPGMWERTL TIGSAGKTFS ATGWKVGWVL GPDHIMKHLR TVHQNSVFHC PTQSQAAVAE SFEREQLLFR QPSSYFVQFP QAMQRCRDHM IRSLQSVGLK PIIPQGSYFL ITDISDFKRK MPDLPGAVDE PYDRRFVKWM IKNKGLVAIP VSIFYSVPHQ KHFDHYIRFC FVKDEATLQA MDEKLRKWKV EL | |
Greater than 90% as determined by SDS-PAGE. |
883 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |