missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kynurenine--oxoglutarate transaminase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Kynurenine--oxoglutarate transaminase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7538-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069250

  • 530.00 € / 100µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001116143.1
Kynurenine--oxoglutarate transaminase 1 Protein
Research Use Only
CCBL1
Recombinant Protein
E. coli
MAKQLQARRL DGIDYNPWVE FVKLASEHDV VNLGQGFPDF PPPDFAVEAF QHAVSGDFML NQYTKTFVII IEPFFDCYEP MTMMAGGRPV FVSLKPGPIQ NGELGSSSNW QLDPMELAGK FTSRTKALVL NTPNNPLGKV FSREELELVA SLCQQHDVVC ITDEVYQWMV YDGHQHISIA SLPGMWERTL TIGSAGKTFS ATGWKVGWVL GPDHIMKHLR TVHQNSVFHC PTQSQAAVAE SFEREQLLFR QPSSYFVQFP QAMQRCRDHM IRSLQSVGLK PIIPQGSYFL ITDISDFKRK MPDLPGAVDE PYDRRFVKWM IKNKGLVAIP VSIFYSVPHQ KHFDHYIRFC FVKDEATLQA MDEKLRKWKV EL
Greater than 90% as determined by SDS-PAGE.
883
100 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions