missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KCND2 Protein

A DNA sequence encoding the Homo sapiens (Human) KCND2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9228-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097871

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_036413.1
KCND2 Protein
Research Use Only
KCND2
Recombinant Protein
Yeast
VSNFSRIYHQ NQRADKRRAQ KKARLARIRA AKSGSANAYM QSKRNGLLSN QLQSSEDEQA FVSKSGSSFE TQHHHLLHCL EKTTNHEFVD EQVFEESCME VATVNRPSSH SPSLSSQQGV TSTCCSRRHK KTFRIPNANV SGSHQGSIQE LSTIQIRCVE RTPLSNSRSS LNAKMEECVK LNCEQPYVTT AIISIPTPPV TTPEGDDRPE SPEYSGGNIV RVSAL
Greater than 90% as determined by SDS-PAGE.
3751
1 mg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions