missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ISOC2 Protein
Specifications
Specifications
| Name | ISOC2 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSEQYPQGL GPTVPELGTE GLRPLAKTCF SMVPALQQEL DSRPQLRSVL LCGIEAQACI LNTTLDLLDR GLQVHVVVDA CSSRSQVDRL VALARMRQSG AFLSTSEGLI LQLVGDAVHP QFKEIQKLIK EPAPDSGLLG LFQGQNSLLH |
| Buffer | The ISOC2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?