missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 8 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human Integrin alpha V beta 8 at 2 μg/mL can bind recombinant human Lap with an apparent Kd <0.5 nM.
Specifications
Specifications
| Accession Number | NP_002201.1 (Integrin alpha V) & P26012.1 (Integrin beta 8) |
| For Use With (Application) | Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and MgCl2. |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | M.W. (Observed): 130-155 kDa and 85-100 kDa, reducing conditions; M.W. (theoretical): 110.5 kDa (alpha V subunit), 75.3 kDa (beta 8 subunit) |
| Name | Integrin alpha V beta 8 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha V beta 8 protein Human Integrin alpha V (Phe31-Val992) (N-terminus) Accession # NP_002201.1 His-Pro GGGSGGGS Acidic Tail Human Integrin beta 8 (Glu43-Arg684) (N-terminus) Accession # P26012.1 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction