missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin alpha-IIb Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-IIb, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6239-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051840

  • 645.00 € / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000410.2
Integrin alpha-IIb Protein
Research Use Only
ITGA2B
Recombinant Protein
Yeast
CVPQLQLTAS VTGSPLLVGA DNVLELQMDA ANEGEGAYEA ELAVHLPQGA HYMRALSNVE GFERLICNQK KENETRVVLC ELGNPMKKNA QIGIAMLVSV GNLEEAGESV SFQLQIRSKN SQNPNSKIVL LDVPVRAEAQ VELRGNSFPA SLVVAAEEGE REQNSLDSWG PKVEHTYELH NNGPGTVNGL HLSIHLPGQS QPSDLLYILD IQPQGGLQCF PQPPVNPLKV DWGLPIPSPS PIHPAHHKR
Greater than 90% as determined by SDS-PAGE.
3674
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions