Learn More
R&D Systems™ Recombinant Human Integrin alpha 11 beta 1 Protein
Description
The Recombinant Human Integrin alpha 11 beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha 11 beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 134.6 kDa ( alpha11) & 88.4 (beta1) |
| Name | Integrin alpha 11 beta 1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 11 beta 1 protein, (N-terminus) Human Integrin alpha 11 (Phe23-Pro1141) (Leu524Arg) Accession &Num; Q9UKX5 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |