missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Inositol-trisphosphate 3-kinase B Protein

A DNA sequence encoding the Homo sapiens (Human) Inositol-trisphosphate 3-kinase B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6243-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051870

  • 518.00 € / 100µg

Estimated Shipment: 23-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_002212.3
Inositol-trisphosphate 3-kinase B Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3707
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ITPKB
Recombinant Protein
E. coli
RVEGGSPTLG LLGGSPSAQP GTGNVEAGIP SGRMLEPLPC WDAAKDLKEP QCPPGDRVGV QPGNSRVWQG TMEKAGLAWT RGTGVQSEGT WESQRQDSDA LPSPELLPQD PDKPFLRKAC SPSNIPAVII TDMGTQEDGA LEETQGSPRG NLPLRKLSSS SASSTGFSSS YEDSEEDISS DPERTLDPNS AFLHTLDQQK PRVSKSWRKI KNMVHWSPFV MSFKKKYPWI QLAGHAGSFK AAANGRILKK HCESEQRCLD RLMVDVLRPF VPAYHGDVVK DGERYNQMDD LLADFDSPCV MDCKMGIRTY LEEELTKARK KPSLRKDMYQ KMIEVDPEAP TEEEKAQRAV TKPRYMQWRE TISSTATLGF RIEGIKKEDG TVNRDFKKTK TREQVTEAFR EFTKGNHNIL IAYRDRLKAI RTTLEVSPFF KCHEVIGSSL LFIHDKKEQA KVWMIDFGKT TPLPEGQTLQ HDVPWQEGNR EDGYLSGLNN LVDILTEMSQ DAPLA
Videos
SDS
Documents
Certificates
Promotions

Promotions