missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Immunity-related GTPase family M Protein

A DNA sequence encoding the Homo sapiens (Human) Immunity-related GTPase family M protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6236-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051810

  • 518.00 € / 100µg

Estimated Shipment: 16-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001139277.1
Immunity-related GTPase family M Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
345611
Greater than 80% as determined by SDS-PAGE.
Research Use Only
IRGM
Recombinant Protein
E. coli
MEAMNVEKAS ADGNLPEVIS NIKETLKIVS RTPVNITMAG DSGNGMSTFI SALRNTGHEG KASPPTELVK ATQRCASYFS SHFSNVVLWD LPGTGSATTT LENYLMEMQF NRYDFIMVAS AQFSMNHVML AKTAEDMGKK FYIVWTKLDM DLSTGALPEV QLLQIRENVL ENLQKERVCE Y
Videos
SDS
Documents
Certificates
Promotions

Promotions