missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL6R / CD126 Protein
A DNA sequence encoding the Homo sapiens (Human) IL6R / CD126, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9217-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000556.1 | |
IL6R / CD126 Protein | |
Research Use Only | |
IL6R | |
Recombinant Protein | |
Yeast | |
LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQDS SSVPLP | |
Greater than 90% as determined by SDS-PAGE. |
3570 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |