missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL6R / CD126 Protein

A DNA sequence encoding the Homo sapiens (Human) IL6R / CD126, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9217-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007861

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000556.1
IL6R / CD126 Protein
Research Use Only
IL6R
Recombinant Protein
Yeast
LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQDS SSVPLP
Greater than 90% as determined by SDS-PAGE.
3570
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions