missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HOXA9 Protein
A cDNA sequence encoding the HOXA9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12295-20ug
Additional Details : Weight : 0.01000kg
Specifications
HOXA9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATTGAL GNYYVDSFLL GADAADELSV GRYAPGTLGQ PPRQAATLAE HPDFSPCSFQ SKATVFGASW NPVHAAGANA VPAAVYHHHH HHPYVHPQAP VAAAAPDGRY MRSWLEPTPG ALSFAGLPSS RPYGIKPEPL SARRGDCPTL DTHTLSLTDY ACGSPPVDRE KQPSEGAFSE NNAENESGGD KPPIDPNNPA ANWLHARSTR KKRCPYTKHQ TLELEKEFLF NMYLTRDRRY EVARLLNLTE RQVKIWFQNR RMKMKKINKD RAKDE | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HOXA9 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |