missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Homer protein homolog 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Homer protein homolog 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8433-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044591

  • 2365.00 € / 1mg

Estimated Shipment: 22-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001264006.1
Homer protein homolog 1 Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
9456
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HOMER1
Recombinant Protein
E. coli
MGEQPIFSTR AHVFQIDPNT KKNWVPTSKH AVTVSYFYDS TRNVYRIISL DGSKAIINST ITPNMTFTKT SQKFGQWADS RANTVYGLGF SSEHHLSKFA EKFQEFKEAA RLAKEKSQEK MELTSTPSQE SAGGDLQSPL TPESINGTDD ERTPDVTQNS EPRAEPTQNA LPFSHSSAIS KHWEAELATL KGNNAKLTAA LLESTANVKQ WKQQLAAYQE EAERLHKRVT ELECVSSQAN AVHTHKTELN QTIQELEETL KLKEEEIERL KQEIDNAREL QEQRDSLTQK LQEVEIRNKD LEGQLSDLEQ RLEKSQNEQE AFRNNLKTLL EILDGKIFEL TELRDNLAKL LECS
Videos
SDS
Documents
Certificates
Promotions

Promotions