missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homer protein homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Homer protein homolog 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8433-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001264006.1 | |
Homer protein homolog 1 Protein | |
Research Use Only | |
HOMER1 | |
Recombinant Protein | |
E. coli | |
MGEQPIFSTR AHVFQIDPNT KKNWVPTSKH AVTVSYFYDS TRNVYRIISL DGSKAIINST ITPNMTFTKT SQKFGQWADS RANTVYGLGF SSEHHLSKFA EKFQEFKEAA RLAKEKSQEK MELTSTPSQE SAGGDLQSPL TPESINGTDD ERTPDVTQNS EPRAEPTQNA LPFSHSSAIS KHWEAELATL KGNNAKLTAA LLESTANVKQ WKQQLAAYQE EAERLHKRVT ELECVSSQAN AVHTHKTELN QTIQELEETL KLKEEEIERL KQEIDNAREL QEQRDSLTQK LQEVEIRNKD LEGQLSDLEQ RLEKSQNEQE AFRNNLKTLL EILDGKIFEL TELRDNLAKL LECS | |
Greater than 90% as determined by SDS-PAGE. |
9456 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |