missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DOB Protein
A cDNA sequence encoding the HLA-DOB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12275-20ug
Additional Details : Weight : 0.01000kg
Specifications
HLA-DOB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGTDSPED FVIQAKADCY FTNGTEKVQF VVRFIFNLEE YVRFDSDVGM FVALTKLGQP DAEQWNSRLD LLERSRQAVD GVCRHNYRLG APFTVGRKVQ PEVTVYPERT PLLHQHNLLH CSVTGFYPGD IKIKWFLNGQ EERAGVMSTG PIRNGDWTFQ TVVMLEMTPE LGHVYTCLVD HSSLLSPVSV EWRAQSEYSW RK | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HLA-DOB solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |