missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone deacetylase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6150-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051180

  • 476.00 € / 100µg
Estimated Shipment: 27-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004955.2
Histone deacetylase 1 Protein
Research Use Only
HDAC1
Recombinant Protein
E. coli
MAQTQGTRRK VCYYYDGDVG NYYYGQGHPM KPHRIRMTHN LLLNYGLYRK MEIYRPHKAN AEEMTKYHSD DYIKFLRSIR PDNMSEYSKQ MQRFNVGEDC PVFDGLFEFC QLSTGGSVAS AVKLNKQQTD IAVNWAGGLH HAKKSEASGF CYVNDIVLAI LELLKYHQRV LYIDIDIHHG DGVEEAFYTT DRVMTVSFHK YGEYFPGTGD LRDIGAGKGK YYAVNYPLRD GIDDESYEAI FKPVMSKVME MFQPSAVVLQ CGSDSLSGDR LGCFNLTIKG HAKCVEFVKS FNLPMLMLGG GGYTIRNVAR CWTYETAVAL DTEIPNELPY NDYFEYFGPD FKLHISPSNM TNQNTNEYLE KIKQRLFENL RMLPHAPGVQ MQAIPEDAIP EESGDEDEDD PDKRISICSS DKRIACEEEF SDSEEEGEGG RKNSSNFKKA KRVKTEDEKE KDPEEKKEVT EEEKTKEEKP EAKGVKEEVK LA
Greater than 90% as determined by SDS-PAGE.
3065
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions