missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HBG1 Protein

A cDNA sequence encoding the HBG1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12121-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949438

  • 137.00 € / 2µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

Specifications

Specifications

HBG1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGHFTEE DKATITSLWG KVNVEDAGGE TLGRLLVVYP WTQRFFDSFG NLSSASAIMG NPKVKAHGKK VLTSLGDATK HLDDLKGTFA QLSELHCDKL HVDPENFKLL GNVLVTVLAI HFGKEFTPEV QASWQKMVTA VASALSSRYH
Greater than 85% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HBG1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions