missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAUS augmin-like complex subunit 4 Protein
A DNA sequence encoding the Homo sapiens (Human) HAUS augmin-like complex subunit 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8087-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001159741.1 | |
HAUS augmin-like complex subunit 4 Protein | |
Research Use Only | |
HAUS4 | |
Recombinant Protein | |
E. coli | |
MASGDFCSPG EGMEILQQVC SKQLPPCNLS KEDLLQNPYF SKLLLNLSQH VDESGLSLTL AKEQAQAWKE VRLHKTTWLR SEILHRVIQE LLVDYYVKIQ DTNVTSEDKK FHETLEQRLL VTELMRLLGP SQEREIPPLL GLEKADLLEL MPLSEDFVWM RARLQQEVEE QLKKKCFTLL CYYDPNSDAD SETVKAAKVW KLAEVLVGEQ QQCQDAKSQQ KEQMLLLEKK SAAYSQVLLR CLTLLQRLLQ EHRLKTQSEL DRINAQYLEV KCGAMILKLR MEELKILSDT YTVEKVEVHR LIRDRLEGAI HLQEQDMENS RQVLNSYEVL GEEFDRLVKE YTVLKQATEN KRWALQEFSK VYR | |
Greater than 80% as determined by SDS-PAGE. |
54930 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |