missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human H1F0 / Histone H1 Protein
A DNA sequence encoding the Homo sapiens (Human) H1F0 / Histone H1, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_005309.1 |
---|---|
Gene ID (Entrez) | 3005 |
Name | H1F0 / Histone H1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091080
|
enQuireBio™
QP6137-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011090
|
enQuireBio™
QP6137-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001090
|
enQuireBio™
QP6137-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021090
|
enQuireBio™
QP6137-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031090
|
enQuireBio™
QP6137-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041090
|
enQuireBio™
QP6137-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005309.1 | |
H1F0 / Histone H1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3005 | |
Research Use Only | |
H1F0 | |
Recombinant Protein | |
E. coli | |
MTENSTSAP AAKPKRAKAS KKSTDHPKYS DMIVAAIQAE KNRAGSSRQS IQKYIKSHYK VGENADSQIK LSIKRLVTTG VLKQTKGVGA SGSFRLAKSD EPKKSVAFKK TKKEIKKVAT PKKASKPKKA ASKAPTKKPK ATPVKKAKKK LAATPKKAKK PKTVKAKPVK ASKPKKAKPV KPKAKSSAKR AGKKK | |
Greater than 80% as determined by SDS-PAGE. |