missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Recombinant protein for Human GSK-3 alpha The Recombinant Human GSK-3 alpha Protein is derived from E. coli. The Recombinant Human GSK-3 alpha Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Accession Number | P49840 |
| Concentration | LYOPH |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from endotoxin free water. |
| Gene ID (Entrez) | 2931 |
| Name | GSK-3 alpha Protein |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Reconstitution | Reconstitute in sterile water. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?