missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Granzyme B / GZMB Protein
A DNA sequence encoding the Homo sapiens (Human) Granzyme B / GZMB, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6136-ye-10ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_004122.2 | |
Granzyme B / GZMB Protein | |
Research Use Only | |
GZMB | |
Recombinant Protein | |
Yeast | |
IIGGHEAKPH SRPYMAYLMI WDQKSLKRCG GFLIRDDFVL TAAHCWGSSI NVTLGAHNIK EQEPTQQFIP VKRPIPHPAY NPKNFSNDIM LLQLERKAKR TRAVQPLRLP SNKAQVKPGQ TCSVAGWGQT APLGKHSHTL QEVKMTVQED RKCESDLRHY YDSTIELCVG DPEIKKTSFK GDSGGPLVCN KVAQGIVSYG RNNGMPPRAC TKVSSFVHWI KKTMKRY | |
Greater than 90% as determined by SDS-PAGE. |
3002 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |