missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutaminyl cyclase / QPCT Protein

A DNA sequence encoding the Homo sapiens (Human) Glutaminyl cyclase / QPCT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6564-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054200

  • 518.00 € / 100µg

Estimated Shipment: 23-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_036545.1
Glutaminyl cyclase / QPCT Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
25797
Greater than 90% as determined by SDS-PAGE.
Research Use Only
QPCT
Recombinant Protein
E. coli
VSPSASAWPE EKNYHQPAIL NSSALRQIAE GTSISEMWQN DLQPLLIERY PGSPGSYAAR QHIMQRIQRL QADWVLEIDT FLSQTPYGYR SFSNIISTLN PTAKRHLVLA CHYDSKYFSH WNNRVFVGAT DSAVPCAMML ELARALDKKL LSLKTVSDSK PDLSLQLIFF DGEEAFLHWS PQDSLYGSRH LAAKMASTPH PPGARGTSQL HGMDLLVLLD LIGAPNPTFP NFFPNSARWF ERLQAIEHEL HELGLLKDHS LEGRYFQNYS YGGVIQDDHI PFLRRGVPVL HLIPSPFPEV WHTMDDNEEN LDESTIDNLN KILQVFVLEY LHL
Videos
SDS
Documents
Certificates
Promotions

Promotions