Learn More
enQuireBio™ Recombinant Human Gasdermin-C Protein
A DNA sequence encoding the Homo sapiens (Human) Gasdermin-C, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8195-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16083201
  • 3405.00 € / 1mg
Estimated Shipment: 05-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Specifications
NP_113603.1
Gasdermin-C Protein
Research Use Only
GSDMC
Recombinant Protein
E. coli
MPSMLERISK NLVKEIGSKD LTPVKYLLSA TKLRQFVILR KKKDSRSSFW EQSDYVPVEF SLNDILEPSS SVLETVVTGP FHFSDIMIQK HKADMGVNVG IEVSVSGEAS VDHGCSLEFQ IVTIPSPNLE DFQKRKLLDP EPSFLKECRR RGDNLYVVTE AVELINNTVL YDSSSVNILG KIALWITYGK GQGQGESLRV KKKALTLQKG MVMAYKRKQL VIKEKAILIS DDDEQRTFQD EYEISEMVGY CAARSEGLLP SFHTISPTLF NASSNDMKLK PELFLTQQFL SGHLPKYEQV HILPVGRIEE PFWQNFKHLQ EEVFQKIKTL AQLSKDVQDV MFYSILAMLR DRGALQDLMN MLELDSSGHL DGPGGAILKK LQQDSNHAWF NPKDPILYLL EAIMVLSDFQ HDLLACSMEK RILLQQQELV RSILEPNFRY PWSIPFTLKP ELLAPLQSEG LAITYGLLEE CGLRMELDNP RSTWDVEAKM PLSALYGTLS LLQQLAEA
Greater than 90% as determined by SDS-PAGE.
56169
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers