missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Galectin-9 Protein

Product Code. 18371688
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 18371688

Brand: R&D Systems™ 9064GA050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

E.coli expressed rh Galectin-9 (Catalog # 2045-GA) is also available with high TIM-3 binding affinity!

Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is 0.75-4.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002299
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, EDTA, DTT and Trehalose.
Gene ID (Entrez) 3965
Molecular Weight (g/mol) M.W. (Observed): 42-51 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Galectin-9 protein Met1-Thr323
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Common Name Galectin-9
Gene Symbol LGALS9
Biological Activity Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is typically 0.75-4.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Met1-Thr323
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Galectin-9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.