missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fizzy-related protein homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Fizzy-related protein homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8256-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129669.1 | |
Fizzy-related protein homolog Protein | |
Research Use Only | |
FZR1 | |
Recombinant Protein | |
E. coli | |
MDQDYERRLL RQIVIQNENT MPRVTEMRRT LTPASSPVSS PSKHGDRFIP SRAGANWSVN FHRINENEKS PSQNRKAKDA TSDNGKDGLA YSALLKNELL GAGIEKVQDP QTEDRRLQPS TPEKKGLFTY SLSTKRSSPD DGNDVSPYSL SPVSNKSQKL LRSPRKPTRK ISKIPFKVLD APELQDDFYL NLVDWSSLNV LSVGLGTCVY LWSACTSQVT RLCDLSVEGD SVTSVGWSER GNLVAVGTHK GFVQIWDAAA GKKLSMLEGH TARVGALAWN AEQLSSGSRD RMILQRDIRT PPLQSERRLQ GHRQEVCGLK WSTDHQLLAS GGNDNKLLVW NHSSLSPVQQ YTEHLAAVKA IAWSPHQHGL LASGGGTADR CIRFWNTLTG QPLQCIDTGS QVCNLAWSKH ANELVSTHGY SQNQILVWKY PSLTQVAKLT GHSYRVLYLA MSPDGEAIVT GAGDETLRFW NVFSKTRSTK ESVSVLNLFT RIR | |
Greater than 90% as determined by SDS-PAGE. |
51343 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |