Learn More
enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein
Product Code. 15915819
Click to view available options
Quantity:
1 mg
20 μg
500 μg
Unit Size:
1mg
20µg
500µg
Product Code. 15915819
Brand: enQuireBio™ QP5272500ug
Please to purchase this item. Need a web account? Register with us today!
Recombinant Protein
Specifications
Accession Number NP_061986.1
Gene ID (Entrez) 26291
Name FGF21 / Fibroblast Growth Factor 21 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol FGF21
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MHPIPDSSPL . LQFGGQVRQ R. YLYTDDAQ QT. EAHLEIR EDG. TVGGAA DQSP. ESLLQ LKALK. PGVI QILGVK. TSR FLCQRPD. GA LYGSLHFD. P EACSFRELLL EDGYNVYQS.  EAHGLPLHLP . GNKSPHRDP A. PRGPARFL PL. PGLPPAL PEP. PGILAP QPPD. VGSSD PLSMV. GPSQ GRSPSY. AS
Buffer 10 mM sodium phosphate, 100 mM soldium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.