missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FGF basic/FGF2 Protein, Animal-Free

Product Code. 18735925
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
This item is not returnable. View return policy

Product Code. 18735925

Brand: R&D Systems™ BTFGFBAFL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Intended for preclinical researchers who may transition to GMP FGF basic/FGF2 for their clinical work

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.100-1.00 ng/mL.The specific activity of Recombinant Human FGF basic/FGF2 is >8.0 x 105 IU/mg, which is calibrated against the human FGF basic/FGF2 WHO International Standard (NIBSC code: 90/712).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001997.5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) M.W. (Observed): 18 kDa, under reducing conditions., M.W. (Theroretical): 16 kDa
Quantity 25 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
Conjugate Unconjugated
Reconstitution Reconstitute 25 μg size at 250 μg/mL and others at 500 μg/mL in sterile water.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.