missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-4, Animal-Free Protein
Description
No significant difference between EC50 of reference and test lots
Specifications
Specifications
| Accession Number | P08620.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from HEPES/NaCl/mannitol |
| Gene ID (Entrez) | 2249 |
| Molecular Weight (g/mol) | M.W. (Observed): Monomeric FGF-4 protein only, M.W. (Theroretical): 14 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human FGF-4 protein |
| Storage Requirements | Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?