missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ESAM / Endothelial Cell Adhesion Molecule Protein
A DNA sequence encoding the Homo sapiens (Human) ESAM / Endothelial Cell Adhesion Molecule, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_620411.2 |
---|---|
Gene ID (Entrez) | 90952 |
Name | ESAM / Endothelial Cell Adhesion Molecule Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071841
|
enQuireBio™
QP7960-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091841
|
enQuireBio™
QP7960-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081841
|
enQuireBio™
QP7960-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001851
|
enQuireBio™
QP7960-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011851
|
enQuireBio™
QP7960-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021851
|
enQuireBio™
QP7960-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_620411.2 | |
ESAM / Endothelial Cell Adhesion Molecule Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
90952 | |
Research Use Only | |
ESAM | |
Recombinant Protein | |
E. coli | |
QLQLHLPANR LQAVEGGEVV LPAWYTLHGE VSSSQPWEVP FVMWFFKQKE KEDQVLSYIN GVTTSKPGVS LVYSMPSRNL SLRLEGLQEK DSGPYSCSVN VQDKQGKSRG HSIKTLELNV LVPPAPPSCR LQGVPHVGAN VTLSCQSPRS KPAVQYQWDR QLPSFQTFFA PALDVIRGSL SLTNLSSSMA GVYVCKAHNE VGTAQCNVTL EVSTGPGAA | |
Greater than 90% as determined by SDS-PAGE. |