missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dual specificity protein phosphatase 26 Protein
A DNA sequence encoding the Homo sapiens (Human) Dual specificity protein phosphatase 26, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8061-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001292044.1 | |
Dual specificity protein phosphatase 26 Protein | |
Research Use Only | |
DUSP26 | |
Recombinant Protein | |
E. coli | |
MCPGNWLWAS MTFMARFSRS SSRSPVRTRG TLEEMPTVQH PFLNVFELER LLYTGKTACN HADEVWPGLY LGDQDMANNR RELRRLGITH VLNASHSRWR GTPEAYEGLG IRYLGVEAHD SPAFDMSIHF QTAADFIHRA LSQPGGKILV HCAVGVSRSA TLVLAYLMLY HHLTLVEAIK KVKDHRGIIP NRGFLRQLLA LDRRLRQGLE A | |
Greater than 90% as determined by SDS-PAGE. |
78986 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |